missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Synthase O Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA597485
Additional Details : Weight : 0.01000kg
Description
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.Spécification
ATP Synthase O | |
Polyclonal | |
Unconjugated | |
ATP5PO | |
ATP synthase peripheral stalk subunit OSCP, ATP synthase subunit O, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit, ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit (oligomycin sensitivity conferring protein), ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit, DNA segment, Chr 12, Wayne State University 28, expressed, ATP5O, ATP5PO, ATPO, D12Wsu28e, HMC08D05, human ATP synthase OSCP subunit, oligomycin sensitivity conferral protein, oligomycin sensitivity conferral protein oscp-like protein, oligomycin sensitivity conferring protein, OSCP, Sperm flagella protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
539 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
P48047 | |
ATP5PO | |
Recombinant Human ATP synthase subunit O, mitochondrial protein (24-213AA) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |