missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Synthase O Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5103629
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total ATP5O.
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.Specifications
ATP Synthase O | |
Polyclonal | |
Unconjugated | |
ATP5PO | |
ATP synthase peripheral stalk subunit OSCP; ATP synthase subunit O, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit (oligomycin sensitivity conferring protein); ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; DNA segment, Chr 12, Wayne State University 28, expressed; ATP5O; ATP5PO; ATPO; D12Wsu28e; HMC08D05; human ATP synthase OSCP subunit; oligomycin sensitivity conferral protein; oligomycin sensitivity conferral protein oscp-like protein; oligomycin sensitivity conferring protein; OSCP; Sperm flagella protein 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
192241, 28080, 539 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P48047, Q06647, Q9DB20 | |
ATP5PO | |
A synthesized peptide derived from human ATP5O(Accession P48047), corresponding to amino acid residues V32-Y82. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |