missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1G3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19523-1-AP-150UL
This item is not returnable.
View return policy
Description
ATPase, H+ transporting, lysosomal (vacuolar proton pump) subunit G3,ATPase, H+ transporting, lysosomal 13kD, V1 subunit G,ATPase, H+ transporting, lysosomal, V1 subunit G3,OTTHUMP00000033686,V-ATPase 13 kDa subunit 3,V-ATPase G subunit 3,V-ATPase G3 subunitSpecifications
| ATP6V1G3 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1G3 | |
| ATP6G3, ATP6V1G3, V ATPase 13 kDa subunit 3, V ATPase subunit G 3, Vma10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 127124, 338375 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BMC1, Q96LB4 | |
| Atp6v1g3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction