missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRAID Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25548-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes all the three isoforms (20-24 kDa, 16-19 kDa and 28-30 kDa).
Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.Specifications
| ATRAID | |
| Polyclonal | |
| Unconjugated | |
| ATRAID | |
| Apoptosis related protein 3, APR 3, APR 3, APR3, C2orf28, HSPC013, p18, PRO240, UNQ214/PRO240 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 381629, 51374 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6PGD0, Q6UW56 | |
| ATRAID | |
| ATRAID Fusion Protein Ag22168 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction