Learn More
ATRAID Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25548-1-AP-20UL
 View more versions of this product
Product Code. 16864115
  • 145.00€ / 20µL
Description
This antibody recognizes all the three isoforms (20-24 kDa, 16-19 kDa and 28-30 kDa).
Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.
Specifications
Specifications
ATRAID
Polyclonal
Unconjugated
ATRAID
Apoptosis related protein 3, APR 3, APR 3, APR3, C2orf28, HSPC013, p18, PRO240, UNQ214/PRO240
Rabbit
Antigen Affinity Chromatography
RUO
381629, 51374
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6PGD0, Q6UW56
ATRAID
ATRAID Fusion Protein Ag22168
20 μL
Primary
Human, Mouse