missing translation for 'onlineSavingsMsg'
Learn More

Atropine Sulfate Monohydrate, TRC

Product Code. 30011704
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011704 25 g 25g
30005121 5 g 5g
18041410 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011704 Supplier Toronto Research Chemicals Supplier No. TRCA79462525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5908-99-6
Molecular Formula 2C17H23NO3.H2O4S.H2O
Molecular Weight (g/mol) 694.83
Synonym Atropine Sulphate, Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester sulfate, hydrate (2:1:1), 1αH,5αH-Tropan-3α-ol, (±)-tropate (ester), sulfate (2:1), hemihydrate (8CI), Benzeneacetic acid, α-(hydroxymethyl)- (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, sulfate (2:1) (salt), monohydrate (9CI), Benzeneacetic acid, α-(hydroxymethyl)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester endo-(±)-, sulfate (2:1) (salt), monohydrate, Atropine sulfate hydrate, Atropine sulfate monohydrate
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
SMILES O.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3.CN4[C@@H]5CC[C@H]4C[C@H](C5)OC(=O)[C@@H](CO)c6ccccc6.OS(=O)(=O)O

Specifications

Analyte or Component Names Atropine sulfate hydrate
Shipping Condition Room Temperature
InChI Formula InChI=1S/2C17H23NO3.H2O4S.H2O/c2*1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12;1-5(2,3)4;/h2*2-6,13-16,19H,7-11H2,1H3;(H2,1,2,3,4);1H2/t2*13-,14+,15+,16-;;/m00../s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 g
Formula Weight 694.3135
Chemical Name or Material Atropine sulfate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.