missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRX / RAD54 Mouse, Clone: 39f, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 510.00€
Specifications
Antigen | ATRX / RAD54 |
---|---|
Clone | 39f |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930688
|
EnquireBio
546-MSM1-P1 |
100 μg |
510.00€
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920688
|
EnquireBio
546-MSM1-P0 |
20 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
ATRX / RAD54 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Recombinant human ATRX protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
280kDa |
39f | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
ATRX | |
IgG1 κ | |
Protein A/G Purified | |
Nuclear | |
Raji, HeLa or A431 Cells. Pancreas, Adrenal, Ovary, Kidney, Prostate or Stomach. |