Learn More
Heidolph™ Attachment for Test Tubes
Product Code. 10049130 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
Test Tubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10049130 Test Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10049130 Supplier Heidolph™ Supplier No. 5490100000
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Reax top and Reax control
For up to 10 test tubes with outer diameter of 10 mm and length up to 60 mm
TRUSTED_SUSTAINABILITY
Specifications
Holds Test Tubes
Tube Size 10 mm dia. x 60 mm L
For Use With (Equipment) Up to 10 test tubes with 10 mm diameter up to 60 mm length
No. of Positions 10