Learn More
Heidolph™ Attachment for Bottles
Product Code. 11711649 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
0.5 L bottles
1 L bottles
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11711649 1 L bottles Pack of 4
11721649 0.5 L bottles Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11711649 Supplier Heidolph™ Supplier No. 5492600000
  • ONLINE EXCLUSIVE PRICE
    69.50€
    63.00€ / Pack of 4
    Save 6.50€
    9% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Reax 20/4, 20/8 and 20/12 shakers
Allows for the use of (4) 0.5 or 1 L bottles
TRUSTED_SUSTAINABILITY
Specifications
Holds 1 L bottles
For Use With (Equipment) Reax 20/4, 20/8 and 20/12
No. of Positions 4
Packaging Quantity 4 Pk.