missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Attractin Monoclonal Antibody (750003)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561790
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Multiple transcript variants encoding different isoforms exist for this gene. One of the isoforms is a membrane-bound protein with sequence similarity to the mouse mahogany protein, a receptor involved in controlling obesity. The other isoform is a secreted protein involved in the initial immune cell clustering during inflammatory responses that may regulate the chemotactic activity of chemokines.
Specifications
| Attractin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O75882 | |
| ATRN | |
| Chinese hamster ovary cell line CHO-derived recombinant human Attractin, Ala84-Gln1272. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry | |
| 750003 | |
| Unconjugated | |
| ATRN | |
| ATRN; atrn {ECO:0000312; Attractin; attractin-2; AW558010; DPPT-L; KIAA0548; mahogany; mahogany homolog; mahogany protein; membrane attractin; mg; MGC126754; Mgca; mKIAA0548; Protein mahogany; protein zitter; RGD:69063} | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction