Learn More
Invitrogen™ Attune™ NxT XE3 Computer Upgrade Kit
Product Code. 17186358
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17186358 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17186358 Supplier Invitrogen™ Supplier No. A43668
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for use with the Attune NxT Acoustic Focusing Cytometer
  • The Invitrogen Attune NxT XE3 Computer Upgrade kit is designed for use with the Attune NxT Acoustic Focusing Cytometer.
  • It includes an OptiPlex XE3 Minitower (Windows 10 Professional 64-bit operating system, 4th Gen Intel Core I7-8700 processor, 32 GB (2x16GB) RAM, 2 GB AMD Radeon video card, 2x2TB SATA hard drive with 64 MB cache, 16X DVD+/- RW drive, network interface card), Dell keyboard (US English), Dell mouse, USB 3.0 Card, USB 3.0 cable, LAN cable, and power cord. Monitor not included.
  • One year from installation for parts, labor, and service travel pursuant to the terms of the Thermofisher Scientific standard limited warranty provisions.
  • One year parts and labor (as provided in written limited warranty statement and accompanying terms in the user manual or other product documentation).
TRUSTED_SUSTAINABILITY
Specifications
Product Line Attune
Content And Storage • OptiPlex XE3 Minitower (Windows 10 Professional 64-bit operating system, 4th Gen Intel Core I7-8700 processor, 32 GB (2x16GB) RAM, 2 GB AMD Radeon video card, 2x2TB SATA hard drive with 64 MB cache, 16X DVD+/- RW drive, network interface card)
• Dell keyboard (US English)
• Dell mouse
• USB 3.0 Card
• USB 3.0 cable
• LAN cable
• Power cord
Monitor not included.
Quantity 1
For Use With (Equipment) Attune NxT Acoustic Focusing Cytometer
Product Type Computer Upgrade Kit