Learn More
Invitrogen™ Attune™ Xenith™ Wash Pump Syringe
Description
The Attune Xenith Wash Pump Syringe is part of an auxiliary pump that moves fluids through the Attune Xenith Flow Cytometer for rinsing and cleaning of the system. To keep your instrument in proper working order, it is important to perform regular maintenance, including changing this syringe according to the maintenance schedule laid out in the Attune Xenith user guide.
Specifications
Specifications
| Quantity | 1 Ea. |
| For Use With (Equipment) | Attune Xenith Flow Cytometer |
| Product Type | Wash Pump Syringe |