missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN2L Rabbit anti-Human, Mouse, Proteintech
Rabbit PolyClonal Antibody
Brand: Proteintech 24822-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an ataxin type 2 related protein of unknown function. This protein is a member of the spinocerebellar ataxia (SCAs) family, which is associated with a complex group of neurodegenerative disorders. Several alternatively spliced transcripts encoding different isoforms have been found for this gene.Specifications
| ATXN2L | |
| Polyclonal | |
| Unconjugated | |
| ATXN2L | |
| A2D, A2LG, A2LP, A2RP, Ataxin 2 domain protein, ataxin 2 like, Ataxin 2 like protein, Ataxin 2 related protein, ATXN2L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11273, 233871 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7TQH0, Q8WWM7 | |
| Atxn2l | |
| ATXN2L Fusion Protein Ag20581 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction