Learn More
ATXN7L1 Monoclonal Antibody (OTI4F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505708S
Description
ATXN7L1 Monoclonal antibody specifically detects ATXN7L1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ATXN7L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15151 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4194 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F6 | |
| Unconjugated | |
| MDM4 | |
| 2810423G08Rik; ataxin 7 like 1; ataxin 7-like 1; ataxin 7-like 4; ataxin-7-like protein 1; Ataxin-7-like protein 4; Atxn7l1; Atxn7l4; KIAA1218; RGD1305730 | |
| Full length human recombinant protein of human MDM4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.