Learn More
AURKA Mouse anti-Human, Unconjugated, Clone: 1F4B10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66757-1-IG-150UL
 View more versions of this product
Product Code. 16858925
  • 410.00€ / 150µL
Description
The AURKA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AURKA. This antibody recognizes human, mouse, rat antigen. The AURKA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
AURKA
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1F4B10
Unconjugated
150 μL
6790
IgG3