missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Aurora B Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB500-185PE
This item is not returnable.
View return policy
Description
Aurora B Polyclonal antibody specifically detects Aurora B in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Aurora B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| PE | |
| AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
| The immunogen recognized by this antibody maps to a region between residue 300 and the C-terminus (residue 344) of human Aurora B kinase using the numbering given in Swiss-Prot entry Q96GD4 (GeneID 9212). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
| 9212 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction