missing translation for 'onlineSavingsMsg'
Learn More

Aurothioglucose 80%, TRC

Product Code. 30134911
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134911 5 mg 5mg
30137777 10 mg 10mg
30006973 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134911 Supplier Toronto Research Chemicals Supplier No. TRCA7947905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12192-57-3
Molecular Formula C6 H11 Au O5 S
Molecular Weight (g/mol) 392.18
Synonym Gold, [1-(thio-κS)-D-glucopyranosato-κO2]-, D-Glucose, 1-thio-, S-gold deriv. (6CI), Gold, (1-thio-D-glucopyranosato)- (8CI), Gold, (1-thio-D-glucopyranosato-O2,S1)-, D-Glucopyranose, 1-thio-, gold complex, [1-(Thio-κS)-D-glucopyranosato-κO2]gold, (D-Glucopyranosylthio)gold, Aureotan, Auromyose, Aurothioglucose, Aurumine, Authron, Brenol, Glucose, 1-thio-, gold salt (1:1), Glysanol B, Gold thioglucose, Gold, (D-glucopyranosylthio)-, Oronol, Solganal, Solganal B
IUPAC Name gold(1+);(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxane-2-thiolate
SMILES OC[C@H]1OC2[S-][Au+][OH][C@@H]2[C@@H](O)[C@@H]1O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H12O5S.Au/c7-1-2-3(8)4(9)5(10)6(12)11-2;/h2-10,12H,1H2;/q;+1/p-1/t2-,3-,4+,5-,6?;/m1./s1
Formula Weight 391.9993
Chemical Name or Material Aurothioglucose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.