Learn More
Poulten Graf™ Autoclavable Glass Syringe
Able to be autoclaved at 134°C. Poulten Graf™ Autoclavable Glass Syringes come with an interchangeable piston.
Brand:  Poulten Graf™ 7 10251
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 10645411
  • 63.90€ / Each
Estimated Shipment: 23-05-2024
to see stock.
Description
Description
  • Complies with CE EN 46001 standards
  • Constructed from Durobax™ glass
  • With permanent amber graduation
  • Acid and alkali resistant
Specifications
Specifications
Autoclavable
Syringe
100 mL
Luer Tip
1/Ea.