missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUTS2 Antibody (CL7084) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76530JF525
This item is not returnable.
View return policy
Description
AUTS2 Monoclonal antibody specifically detects AUTS2 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| AUTS2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140 | |
| This antibody was developed using a synthetic peptide derived from Q8WXX7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL7084 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 26053 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction