missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PLZF Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB8395-SP
This item is not returnable.
View return policy
Description
PLZF Monoclonal specifically detects PLZF in Mouse samples. It is validated for Western Blot.
Specifications
| PLZF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_001028496 | |
| ZBTB16 | |
| E. coli-derived recombinant mouse PLZF Met1-Gln254 Accession # NP_001028496 | |
| 25 μg | |
| Primary | |
| Detects mouse PLZF in direct ELISA and Western Blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 816416 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLZFzinc finger and BTB domain-containing protein 16, ZBTB16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145, ZNF145Promyelocytic leukemia zinc finger protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7704 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction