missing translation for 'onlineSavingsMsg'
Learn More

Avian Influenza A H7N9 Neuraminidase Antibody - (A/Shanghai/02/2013) - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-41278-0.025mg

 View more versions of this product

Product Code. 18669437

  • 168.00€ / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Avian Influenza A H7N9 Neuraminidase Polyclonal specifically detects Avian Influenza A H7N9 Neuraminidase in Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Avian Influenza A H7N9 Neuraminidase
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
Rabbit
Affinity Purified
RUO
virus
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
ELISA
1 mg/mL
ELISA 1 μg/mL
EC=3.2.1.18, Neuraminidase
Antibody was raised against a synthetic peptide corresponding to 11 amino acids near the amino terminus of the H7N9 [Influenza A virus (A/Shanghai/02/2013(H7N9))] Neuraminidase protein. The immunogen is located within amino acids 50 - 100 of Avian Influenza A H7N9 Neuraminidase. Amino Acid Squence: QMEERTSRNFN
0.025 mg
Primary
Virus
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.