missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Avidin/Biotin Blocking Kit

Product Code. 15918982
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 150
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15918982 1 Kit Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15918982 Supplier Invitrogen™ Supplier No. 004303

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Avidin/Biotin Blocking Kit

This kit is used to block endogenous biotin or biotin-binding activity in tissue sections. The kit contains 18 mL each of Avidin and Biotin solutions.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Description Avidin/Biotin Blocking Kit, Blocking/Background Suppression Reagent Type, For Cellular Imaging and to Block Endogenous Biotin or Biotin-Binding Activity in Tissue Sections
Includes 18mL Each of avidin and biotin solutions
Quantity 1 Kit
Regulatory Status General purpose reagent, For laboratory use
Content And Storage 2°C to 8°C
For Use With (Application) Cell analysis, cellular imaging, immunohistochemistry (IHC), IHC epitope retrieval, blocking and quenching
Product Type Avidin/Biotin Blocking Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.