missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AWAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49395-25ul
This item is not returnable.
View return policy
Description
AWAT1 Polyclonal antibody specifically detects AWAT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AWAT1 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| acyl-CoA wax alcohol acyltransferase 1, DGA2, DGAT2L3, diacyl-glycerol acyltransferase 2, Diacylglycerol acyltransferase 2, diacylglycerol O-acyltransferase 2-like 3, Diacylglycerol O-acyltransferase 2-like protein 3, EC 2.3.1.75, Long-chain-alcohol O-fatty-acyltransferase 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: WVRNWCVWTHIRDYFPITILKTKDLSPEHNYLMGVHPHGLLTFGAFCNFCT | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 158833 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction