Learn More
Axygen™ 1000 μL Universal Pipetter Tips: Bevelled
GREENER_CHOICE
Product Code. 12176870 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Blue
Clear
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 5000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Color Format unitSize
12176870 Non-sterile Blue Bulk Pack of 1000
12551308 Non-sterile Blue Bulk Pack of 5000
12679535 Non-sterile Blue Rack Pack of 1000
12693706 Sterile Blue Rack Pack of 1000
12186870 Non-sterile Clear Bulk Pack of 1000
12561308 Non-sterile Clear Bulk Pack of 5000
12697215 Non-sterile Clear Rack Pack of 1000
12683706 Sterile Clear Rack Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12176870 Supplier Axygen™ Supplier No. T1000B
Powering Proteomics
Use promo code 28003
View all promotions
In Stock
Add to basket
Edge
Add to basket
Compatible with most popular brands of pipettes
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 100 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Blue
Autoclavable Autoclavable
Model T-1000-B
Certifications/Compliance EN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Quality Control Tested
Clean Claims DNase, RNase, and Pyrogen free
Format Bulk
Tip Style Beveled, Universal
Packaging 1,000 Tips/Pk., 5 Packs/Cs.
Quantity 1000 Tips/Pk., 5 Pk./Cs.
Product Type Pipette Tips
For Use With (Equipment) Universal Fit
Material Polypropylene