Learn More
Axygen™ Gel Cutting Tips: 1.1 x 4 mm
Universal fit for research grade pipettes.
Brand:  Axygen™ TGL-1140-R
Additional Details : Weight : 6.35000kg
 View more versions of this product
Product Code. 12538017
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Axygen™ Gel Cutting Tips are designed to remove bands from agarose gel. These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 μL pipettors
  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Nonsterile
Specifications
Specifications
1000 μL
Clear
DNase, RNase Free
48 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
Polypropylene
Non-sterile
1.1 x 4 mm
Rack
Yes
2, 5μL