missing translation for 'onlineSavingsMsg'
Learn More

Aza-12-crown-4, 99.5+%, Thermo Scientific™

Product Code. 10591881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10591881 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10591881 Supplier Thermo Scientific Acros Supplier No. 277382500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 41775-76-2
Melting Point 53°C to 58°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H18NO3
MDL Number MFCD00134269
Quantity 250 mg
Synonym 1-aza-12-crown 4-ether, aza-12-crown-4, acmc-209spc, 1-aza-12-crown4-ether, 1-aza-4,7,10-trioxacyclododecane, 1,4,7-trioxa-10-aza cyclododecane, 1-aza-12-crown
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,chloroform,acetone
InChI Key QNSRHBOZQLXYNV-UHFFFAOYSA-O
SMILES C1COCCOCCOCC[NH2+]1
IUPAC Name 1,4,7-trioxa-10-azacyclododecane
Molecular Weight (g/mol) 176.24
PubChem CID 545817
Formula Weight 175.23
Percent Purity 99.5+%
Physical Form Crystals, Needles or Flakes
Chemical Name or Material Aza-12-crown-4, 99.5+%
Show More Show Less
compliance-icons
Product Identifier
  • Aza-12-crown-4
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.