missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J62314.03

Quantity 1g

 View more versions of this product

Product Code. 15444719

  • 113.00€ / 1g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in Dichloromethane and dimethyl sulfoxide.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

446-86-6
277.262
LMEKQMALGUDUQG-UHFFFAOYSA-N
2265
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
C9H7N7O2S
MFCD00069203
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
CHEBI:2948
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]

Specifications

446-86-6
MFCD00069203
14,902
Soluble in Dichloromethane and dimethyl sulfoxide.
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]
277.262
CHEBI:2948
Azathioprine
C9H7N7O2S
1g
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
LMEKQMALGUDUQG-UHFFFAOYSA-N
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
2265
277.26
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.