missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine, Thermo Scientific™

113.00€ - 908.00€

Chemical Identifiers

CAS 446-86-6
Molecular Formula C9H7N7O2S
Molecular Weight (g/mol) 277.262
MDL Number MFCD00069203
InChI Key LMEKQMALGUDUQG-UHFFFAOYSA-N
Synonym azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
PubChem CID 2265
ChEBI CHEBI:2948
IUPAC Name 6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
SMILES CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15444719
SDS View Documents
Thermo Scientific Alfa Aesar
J62314.03
1g
113.00€
1g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15454719
SDS View Documents
Thermo Scientific Alfa Aesar
J62314.06
5g
419.00€
5g
View alternative products
Save Up to 
»
Estimated Shipment: 01-10-2026
Add to basket
 
 Restricted Item
15464719
SDS View Documents
Thermo Scientific Alfa Aesar
J62314.09
10g
908.00€
10g
View alternative products
Save Up to 
»
Estimated Shipment: 01-10-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in Dichloromethane and dimethyl sulfoxide.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
Specifications

Chemical Identifiers

446-86-6
277.262
LMEKQMALGUDUQG-UHFFFAOYSA-N
2265
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
C9H7N7O2S
MFCD00069203
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
CHEBI:2948
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]

Specifications

446-86-6
MFCD00069203
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
LMEKQMALGUDUQG-UHFFFAOYSA-N
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
2265
277.26
C9H7N7O2S
14,902
Soluble in Dichloromethane and dimethyl sulfoxide.
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]
277.262
CHEBI:2948
Azathioprine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302-H315-H319-H335
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H350

missing translation for 'einecsNumber' : 207-175-4

missing translation for 'rtecsNumber' : UO8925000

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.