missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine

Product Code. 15464719
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464719 10 g 10g
15444719 1 g 1g
15454719 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15464719 Supplier Thermo Scientific Alfa Aesar Supplier No. J62314.09
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 446-86-6 | C9H7N7O2S | 277.262 g/mol

Azathioprine is an immunosuppressive purine antimetabolite. The mechanism of action of azathioprine is as a nucleic acid synthesis inhibitor. It has shown promise in treatment of alopecia areata (autoimmune hair loss), with no difference in effectiveness between genders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Dichloromethane and dimethyl sulfoxide.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-86-6
Molecular Formula C9H7N7O2S
Molecular Weight (g/mol) 277.262
MDL Number MFCD00069203
InChI Key LMEKQMALGUDUQG-UHFFFAOYSA-N
Synonym azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
PubChem CID 2265
ChEBI CHEBI:2948
IUPAC Name 6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
SMILES CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 10 g
Merck Index 14,902
Solubility Information Soluble in Dichloromethane and dimethyl sulfoxide.
Formula Weight 277.26
Physical Form Crystalline Powder
Chemical Name or Material Azathioprine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.