Learn More
AZD2014, TRC
Product Code. 30179588
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179588 100 mg 100mg
30179266 50 mg 50mg
30179421 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179588 Supplier Toronto Research Chemicals Supplier No. TRCA808115100MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1009298-59-2
Molecular Formula C25H30N6O3
Molecular Weight (g/mol) 462.54
Synonym AZD2014, Vistusertib
IUPAC Name 3-[2,4-bis[(3S)-3-methylmorpholin-4-yl]pyrido[2,3-d]pyrimidin-7-yl]-N-methylbenzamide
SMILES CNC(=O)c1cccc(c1)c2ccc3c(nc(nc3n2)N4CCOC[C@@H]4C)N5CCOC[C@@H]5C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C25H30N6O3/c1-16-14-33-11-9-30(16)23-20-7-8-21(18-5-4-6-19(13-18)24(32)26-3)27-22(20)28-25(29-23)31-10-12-34-15-17(31)2/h4-8,13,16-17H,9-12,14-15H2,1-3H3,(H,26,32)/t16-,17-/m0/s1
Formula Weight 462.2379
Chemical Name or Material 3-[2,4-Bis[(3S)-3-methyl-4-morpholinyl]pyrido[2,3-d]pyrimidin-7-yl]-N-methylbenzamide