missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZI2/NAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15042-1-AP-20UL
This item is not returnable.
View return policy
Description
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).Specifications
| AZI2/NAP1 | |
| Polyclonal | |
| Unconjugated | |
| AZI2 | |
| 5 azacytidine induced 2, AZ2, AZI2, Nak associated protein 1, NAP1, TILP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64343 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H6S1 | |
| AZI2 | |
| AZI2/NAP1 Fusion Protein Ag6892 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction