Learn More
Azlon™ Octagonal Magnetic Stirrer Bar
For stirring applications. Azlon™ Octagonal Magnetic Stirrer Bars are available in multiple lengths.
Brand:  Azlon SWN714
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10115924
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • With central pivot ring
  • Shape gives increased turbulence at low speeds
Specifications
Specifications
10 mm
Central pivot ring
-200°C to 260°C
51 mm
PTFE
Magnetic Stirrer Bar