Learn More
Azlon™ Octagonal Magnetic Stirrer Bar
For stirring applications. Azlon™ Octagonal Magnetic Stirrer Bars are available in multiple lengths.
Brand: Azlon SWN708
This item is not returnable.
View return policy
Description
- With central pivot ring
- Shape gives increased turbulence at low speeds
Specifications
Autoclavable | |
Central pivot ring | |
8 mm | |
Magnet | |
Octagon |
0.51 Lin. | |
0.51 in. | |
13 mm | |
2/Pk. |