Learn More
Azlon™ Octagonal Magnetic Stirrer Bar
For stirring applications. Azlon™ Octagonal Magnetic Stirrer Bars are available in multiple lengths.
Brand: Azlon SWN712
Additional Details : Weight : 0.10000kg
Description
- With central pivot ring
- Shape gives increased turbulence at low speeds
Specifications
Central pivot ring | |
1.49 in. | |
1.49 Lin. | |
2/Pk. | |
Yes | |
Octagon |
8 mm | |
PTFE | |
Stir Bar | |
Magnet | |
38 mm |