Learn More
Azlon™ Octagonal Magnetic Stirrer Bar
For stirring applications. Azlon™ Octagonal Magnetic Stirrer Bars are available in multiple lengths.
Brand: Azlon SWN710
Additional Details : Weight : 0.00990kg
Description
- With central pivot ring
- Shape gives increased turbulence at low speeds
Specifications
Central pivot ring | |
0.98 in. | |
2/Pk. | |
Yes | |
Octagon |
8 mm | |
0.98 Lin. | |
Magnet | |
25 mm |