Learn More
Azlon™ Octagonal Magnetic Stirrer Bar
For stirring applications. Azlon™ Octagonal Magnetic Stirrer Bars are available in multiple lengths.
Brand: Azlon SWN7010
Additional Details : Weight : 0.02000kg
Description
- With central pivot ring
- Shape gives increased turbulence at low speeds
Specifications
10 mm | |
2 | |
Magnet | |
38 mm |