Learn More
Azlon™ Polystyrene Desiccator Cabinet
Product Code. 11313614
Change view
Click to view available options
Color:
Clear
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11313614 Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11313614 Supplier Azlon Supplier No. DWA301
  • ONLINE EXCLUSIVE PRICE
    310.20€
    230.64€ / Pack of 1
    Save 79.56€
    25% Off
Estimated Shipment: 22-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from clear, high impact polystrene for strength and visibility. Azlon™ Polystryrene Dessiccator Cabinets provide moisture and dust free storage of humidity sensitive items.
  • Provides moisture-proof storage
  • Clear high impact polystyrene cabinet
  • Space saving stacking feature
  • Airtight door with foam rubber gasket
  • Supplied with sealed jar of silica gel
Specifications
Color Clear
Material Polystyrene
Includes Sealed jar of silica gel
Dimensions (L x W) 225 x 200 mm
Length (Metric) 225 mm
Width (Metric) 200 mm
Height (Metric) 168 mm