missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZM 475271
Description
Src tyrosine kinase inhibitor. Reduces tumor size, vascularity and metastasis, and increases apoptosis in human pancreatic cells grown in nude mice. Also exhibits antiangiogenic activity in vitro and in vivo. Sensitizes tumor cells to the cytotoxic effects of gemcitabine (Cat. No. 3259).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 442.94 |
| Purity | >99% |
| Molecular Weight (g/mol) | 442.944 |
| Chemical Name or Material | AZM 475271 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?