missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Azurocidin Monoclonal Antibody (D1), Invitrogen™

Mouse Monoclonal Antibody
180.00€ - 453.00€
Specifications
| Antigen | Azurocidin |
|---|---|
| Clone | D1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17654396
|
Invitrogen™
MA543941 |
20 μL |
180.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17644396
![]() |
Invitrogen™
MA543940 |
100 μL |
453.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Azurocidin, also called heparin-binding protein (HBP), is an inactive homolog of serine proteinases residing in granulocytes. Initially it was thought to have only activity against gram-negative bacteria. Later it was also found to act against gram-positive bacteria and C. albicans. Azurocidin is considered a family member of polymorphonuclear leukocytes- derived antimicrobial proteins like defensins, LL-37 and lysozyme. Azurocidin has been recognized as a player in the activation and modulation of the immune response and may act to alarm the immune system. The cationic domain at one side of the protein is essential for its antimicrobial activity. It is stored in azurophil granules as well as secretory granules and as a result partly released at early stage of extravasation. Azurocidin is involved in chemotaxis and activation of monocytes, cytokine release and phagocytosis leading to more efficient bacterial clearance. The primary targeted environment of azurocidin are cells in the bloodstream, that is the endothelial lining, and the extravascular surroundings. The interaction of azurocidin with leucocytes is mediated via beta 2-intergrins. High plasma levels of azurocidin might help with the identification of patients which are at risk in developing sepsis.Specifications
| Azurocidin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| AZAMP; AZU; AZU1; Azurocidin; azurocidin 1; CAP37; cationic antimicrobial protein 37; cationic antimicrobial protein CAP37; HBP; Heparin-binding protein; hHBP; HUMAZUR; NAZC; neutrophil azurocidin; pHBP | |
| AZU1 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| D1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P20160 | |
| 566 | |
| Pro23-Gly247 (Accession P20160), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| AZU1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title