missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ B-Raf (V600E) Recombinant Rabbit Monoclonal Antibody (4R4T5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557887

 View more versions of this product

Product Code. 30284381

  • 176.00€ / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: KTRHVNILLF MGYSTKPQLA IVTQWCEGSS LYHHLHIIET KFEMIKLIDI ARQTAQGMDY LHAKSIIHRD LKSNNIFLHE DLTVKIGDFG LATVKSRWSG.

This gene, B-RAF/BRAF, encodes a protein belonging to the RAF family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERK signaling pathway, which affects cell division, differentiation, and secretion. Mutations in this gene, most commonly the V600E mutation, are the most frequently identified cancer-causing mutations in melanoma, and have been identified in various other cancers as well, including non-Hodgkin lymphoma, colorectal cancer, thyroid carcinoma, non-small cell lung carcinoma, hairy cell leukemia and adenocarcinoma of lung. Mutations in this gene are also associated with cardiofaciocutaneous, Noonan, and Costello syndromes, which exhibit overlapping phenotypes. A pseudogene of this gene has been identified on the X chromosome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

B-Raf (V600E)
Recombinant Monoclonal
1.79 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P15056
Rabbit
Protein A
RUO
673
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
4R4T5
Unconjugated
BRAF
BRAF
A synthetic peptide corresponding to a sequence within amino acids 507-606 of B-Raf (NP_004324.2).
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.