Learn More
BOLA B.Safe Air Valve
Product Code. 17883352
Change view
Click to view available options
Quantity:
100 Pk.
50 Pk.
Unit Size:
Pack of 100
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17883352 100 Pk. Pack of 100
17834014 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17883352 Supplier BOLA Supplier No. M50510
PP with connection UNF 1/4 for B.Safe Caps integrated non-return valve with porous PTFE-Membrane
  • B.Safe Air Valve body made of PP with connection (thread UNF 1/4 28 G) suitable for B.Safe Caps
  • Lid with aeration opening and Luerlock connector, integrated non-return valve with upstream porous PTFE membrane (0.001 mm) including clip-on service life indicator
  • Packing Unit pcs. 50 and 100
  • Service life months 6
  • Porous PTFE Membrane for pressure compensation and protection from contamination, including clip-on service life indicator
Specifications
Length (Metric) 0.001 mm
Includes 6 Service Life Months
Material PTFE
Type Valve
Quantity 100 Pk.