missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B7-H3 Recombinant Rabbit Monoclonal Antibody (23GB5035), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
217.00€ - 590.00€
Specifications
| Antigen | B7-H3 |
|---|---|
| Clone | 23GB5035 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088239
![]() |
Invitrogen™
MA553846 |
100 μL |
590.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089229
|
Invitrogen™
MA553847 |
20 μL |
217.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-H3 Recombinant Monoclonal antibody specifically detects B7-H3 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| B7-H3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5ZPR3 | |
| 80381 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5035 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 | |
| A synthesized peptide derived from human CD276 (1-534AA). | |
| Primary | |
| -20°C | |
| CD276 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title