Learn More
Bode Chemie™ Bacillol AF™ Surface Disinfectant
Product Code. 10320101
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10320101 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10320101 Supplier Bode Chemie™ Supplier No. 9802174
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Bode Chemie™ Bacillol AF™ Surface Disinfectant is for areas that require quick exposure and drying times, and residue free drying.
  • Quick exposure time
  • Quick drying time
  • Residue free
Specifications
Product Type Bacillol AF Disinfectant
Form Liquid
pH 6
Appearance Colorless
Certifications/Compliance CE, MDD, VAH
Composition Active ingredients: Propan-1-ol 400 mg/g, propan-2-ol 250 mg/g, Ethanol 47 mg/g
Container Type Canister
For Use With (Application) For Areas That Require Quick Exposure and Drying Times and Residue Free Drying
Volume (Metric) 5 L