Learn More
Bacitracin (>80%), TRC
Product Code. 30138741
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138741 25 g 25g
30135138 10 g 10g
30006798 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30138741 Supplier Toronto Research Chemicals Supplier No. TRCB10650025G
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1405-87-4
Molecular Formula C66 H103 N17 O16 S . 3 C65 H101 N17 O16 S
Molecular Weight (g/mol) 1155.4
Synonym Altracin, Ayfivin, Baci-Jel, Baciguent, Bacillichin, Bacilliquin, Bacitek Ointment, Fortracin, Parentracin, Penitracin, Topitracin, Tropitracin, Zutracin
IUPAC Name (4R)-4-[[(2S)-2-[[(4R)-2-[(1S,2S)-1-amino-2-methylbutyl]-4,5-dihydro-1,3-thiazole-4-carbonyl]amino]-4-methylpentanoyl]amino]-5-[[(2S)-1-[[(3S,6R,9S,12R,15S,18R,21S)-3-(2-amino-2-oxoethyl)-18-(3-aminopropyl)-12-benzyl-15-[(2S)-butan-2-yl]-6-(carboxymethyl)-9-(1H-imidazol-4-ylmethyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclopentacos-21-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-5-oxopentanoic acid;(4R)-4-[[(2S)-2-[[(4R)-2-[(1S,2S)-1-amino-2-methylbutyl]-4,5-dihydro-1,3-thiazole-4-carbonyl]amino]-4-methylpentanoyl]amino]-5-[[(2S,3S)-1-[[(3S,6R,9S,12R,15S,18R,21S)-3-(2-amino-2-oxoethyl)-18-(3-aminopropyl)-12-benzyl-15-[(2S)-butan-2-yl]-6-(carboxymethyl)-9-(1H-imidazol-4-ylmethyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclopentacos-21-yl]amino]-3-methyl-1-oxopentan-2-yl]amino]-5-oxopentanoic acid;(4R)-4-[[(2S)-2-[[(4R)-2-[(1S,2S)-1-amino-2-methylbutyl]-4,5-dihydro-1,3-thiazole-4-carbonyl]amino]-4-methylpentanoyl]amino]-5-[[(2S,3S)-1-[[(3S,6R,9S,12R,15S,18R,21S)-3-(2-amino-2-oxoethyl)-18-(3-aminopropyl)-12-benzyl-6-(carboxymethyl)-9-(1H-imidazol-4-ylmethyl)-2,5,8,11,14,17,20-heptaoxo-15-propan-2-yl-1,4,7,10,13,16,19-heptazacyclopentacos-21-yl]amino]-3-methyl-1-oxopentan-2-yl]amino]-5-oxopentanoic acid;(4R)-4-[[(2S)-2-[[(4R)-2-[(1S)-1-amino-2-methylpropyl]-4,5-dihydro-1,3-thiazole-4-carbonyl]amino]-4-methylpentanoyl]amino]-5-[[(2S,3S)-1-[[(3S,6R,9S,12R,15S,18R,21S)-3-(2-amino-2-oxoethyl)-18-(3-aminopropyl)-12-benzyl-15-[(2S)-butan-2-yl]-6-(carboxymethyl)-9-(1H-imidazol-4-ylmethyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclopentacos-21-yl]amino]-3-methyl-1-oxopentan-2-yl]amino]-5-oxopentanoic acid
SMILES CC[C@H](C)[C@H](N)C1=N[C@@H](CS1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@H](CCC(=O)O)C(=O)N[C@@H](C(C)C)C(=O)N[C@H]2CCCCNC(=O)[C@H](CC(=O)N)NC(=O)[C@@H](CC(=O)O)NC(=O)[C@H](Cc3c[nH]cn3)NC(=O)[C@@H](Cc4ccccc4)NC(=O)[C@@H](NC(=O)[C@@H](CCCN)NC2=O)[C@@H](C)CC.CC[C@H](C)[C@H](N)C5=N[C@@H](CS5)C(=O)N[C@@H](CC(C)C)C(=O)N[C@H](CCC(=O)O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@H]6CCCCNC(=O)[C@H](CC(=O)N)NC(=O)[C@@H](CC(=O)O)NC(=O)[C@H](Cc7c[nH]cn7)NC(=O)[C@@H](Cc8ccccc8)NC(=O)[C@@H](NC(=O)[C@@H](CCCN)NC6=O)C(C)C.CC[C@H](C)[C@H](N)C9=N[C@@H](CS9)C(=O)N[C@@H](CC(C)C)C(=O)N[C@H](CCC(=O)O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@H]%10CCCCNC(=O)[C@H](CC(=O)N)NC(=O)[C@@H](CC(=O)O)NC(=O)[C@H](Cc%11c[nH]cn%11)NC(=O)[C@@H](Cc%12ccccc%12)NC(=O)[C@@H](NC(=O)[C@@H](CCCN)NC%10=O)[C@@H](C)CC.CC[C@H](C)[C@H](NC(=O)[C@@H](CCC(=O)O)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H]%13CSC(=N%13)[C@@H](N)C(C)C)C(=O)N[C@H]%14CCCCNC(=O)[C@H](CC(=O)N)NC(=O)[C@@H](CC(=O)O)NC(=O)[C@H](Cc%15c[nH]cn%15)NC(=O)[C@@H](Cc%16ccccc%16)NC(=O)[C@@H](NC(=O)[C@@H](CCCN)NC%14=O)[C@@H](C)CC
Especificaciones
Analyte or Component Names Bacitracin
Shipping Condition Room Temperature
InChI Formula InChI=1S/C66H103N17O16S.3C65H101N17O16S/c1-9-35(6)52(69)66-81-48(32-100-66)63(97)76-43(26-34(4)5)59(93)74-42(22-23-50(85)86)58(92)83-53(36(7)10-2)64(98)75-40-20-15-16-25-71-55(89)46(29-49(68)84)78-62(96)47(30-51(87)88)79-61(95)45(28-39-31-70-33-72-39)77-60(94)44(27-38-18-13-12-14-19-38)80-65(99)54(37(8)11-3)82-57(91)41(21-17-24-67)73-56(40)90;1-9-35(7)51(68)65-80-47(31-99-65)62(96)75-42(25-33(3)4)58(92)73-41(21-22-49(84)85)57(91)82-53(36(8)10-2)64(98)74-39-19-14-15-24-70-54(88)45(28-48(67)83)77-61(95)46(29-50(86)87)78-60(94)44(27-38-30-69-32-71-38)76-59(93)43(26-37-17-12-11-13-18-37)79-63(97)52(34(5)6)81-56(90)40(20-16-23-66)72-55(39)89;1-9-35(7)51(68)65-80-47(31-99-65)62(96)75-42(25-33(3)4)58(92)73-41(21-22-49(84)85)57(91)81-52(34(5)6)63(97)74-39-19-14-15-24-70-54(88)45(28-48(67)83)77-61(95)46(29-50(86)87)78-60(94)44(27-38-30-69-32-71-38)76-59(93)43(26-37-17-12-11-13-18-37)79-64(98)53(36(8)10-2)82-56(90)40(20-16-23-66)72-55(39)89;1-9-35(7)52(82-57(91)41(21-22-49(84)85)73-58(92)42(25-33(3)4)75-62(96)47-31-99-65(80-47)51(68)34(5)6)63(97)74-39-19-14-15-24-70-54(88)45(28-48(67)83)77-61(95)46(29-50(86)87)78-60(94)44(27-38-30-69-32-71-38)76-59(93)43(26-37-17-12-11-13-18-37)79-64(98)53(36(8)10-2)81-56(90)40(20-16-23-66)72-55(39)89/h12-14,18-19,31,33-37,40-48,52-54H,9-11,15-17,20-30,32,67,69H2,1-8H3,(H2,68,84)(H,70,72)(H,71,89)(H,73,90)(H,74,93)(H,75,98)(H,76,97)(H,77,94)(H,78,96)(H,79,95)(H,80,99)(H,82,91)(H,83,92)(H,85,86)(H,87,88);11-13,17-18,30,32-36,39-47,51-53H,9-10,14-16,19-29,31,66,68H2,1-8H3,(H2,67,83)(H,69,71)(H,70,88)(H,72,89)(H,73,92)(H,74,98)(H,75,96)(H,76,93)(H,77,95)(H,78,94)(H,79,97)(H,81,90)(H,82,91)(H,84,85)(H,86,87);11-13,17-18,30,32-36,39-47,51-53H,9-10,14-16,19-29,31,66,68H2,1-8H3,(H2,67,83)(H,69,71)(H,70,88)(H,72,89)(H,73,92)(H,74,97)(H,75,96)(H,76,93)(H,77,95)(H,78,94)(H,79,98)(H,81,91)(H,82,90)(H,84,85)(H,86,87);11-13,17-18,30,32-36,39-47,51-53H,9-10,14-16,19-29,31,66,68H2,1-8H3,(H2,67,83)(H,69,71)(H,70,88)(H,72,89)(H,73,92)(H,74,97)(H,75,96)(H,76,93)(H,77,95)(H,78,94)(H,79,98)(H,81,90)(H,82,91)(H,84,85)(H,86,87)/t35-,36-,37-,40-,41+,42+,43-,44+,45-,46-,47+,48-,52-,53-,54-;3*35-,36-,39-,40+,41+,42-,43+,44-,45-,46+,47-,51-,52-,53-/m0000/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 g
Formula Weight 5644.9488
Chemical Name or Material Bacitracin
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.