missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Soytone

Product Code. 16259861
Change view
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16259861 500 g 500g
16249861 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16259861 Supplier Gibco™ Supplier No. 243620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion.

  • An enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion
  • The naturally high concentrations of vitamins and carbohydrates found in soy make Bacto Soytone a nutritious carbohydrate source for those not concerned about the processing step involving an animal-based material
  • This supplement offers a unique nutritional profile as compared to other soy products
  • It can be a valuable alternative to consider for supplementing both mammalian and microbial processes
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Enzymatic Hydrolysate of Soybean meal
Product Line Bacto
Quantity 500 g
Source Animal origin, porcine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.