missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bad Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
197.00€ - 285.00€
Specifications
| Antigen | Bad |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18065885
|
Novus Biologicals
NB100-56080SS |
0.025 mL |
197.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035785
|
Novus Biologicals
NB100-56080 |
0.05 mL |
285.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bad Polyclonal specifically detects Bad in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Bad | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| Whole antisera with 0.02% Sodium Azide | |
| 572 | |
| A full length recombinant protein corresponding to human Bad was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:2000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter | |
| BAD | |
| IgG | |
| Unpurified | |
| Bad |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title