missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (2403C) [DyLight 650], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB1162W
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
BAFFR/TNFRSF13C | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
2403C | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13C | |
Mouse myeloma cell line, NS0-derived human BAFF R/TNFRSF13C, Ser7-Ala71, Accession # Q96RJ3 | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
115650 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction