missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Bafilomycin A1

Product Code. 17140352
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17140352 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17140352 Supplier STEMCELL Technologies Supplier No. 74242
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits autophagy and vacuolar H+-ATPase (V-ATPase)

  • Bafilomycin A1 is a macrolide antibiotic drug and autophagy inhibitor
  • It prevents the acidification of autophagosomes via the inhibition of vacuolar H+-ATPase (V-ATPase; Klionsky et al.)
  • Inhibits the proteolytic degradation associated with autophagy in human mesenchymal stem cells (Oliver et al.)
  • At low concentrations, it inhibits autophagy and promotes apoptotic cell death in pediatric B cell acute lymphoblastic leukemia (B-ALL) cells (Yuan et al.)

Specifications

CAS 88899-55-2
Synonym NSC 381866
Quantity 500 μg
Molecular Formula C35H58O9
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.