missing translation for 'onlineSavingsMsg'
Learn More

Bafilomycin A1, TRC

Product Code. 30136243
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136243 5 mg 5mg
30133388 10 mg 10mg
30011996 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30136243 Supplier Toronto Research Chemicals Supplier No. TRCB1100005MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88899-55-2
Molecular Formula C35H58O9
Molecular Weight (g/mol) 622.83
Synonym 21-O-De(3-carboxy-1-oxo-2-propenyl)-2-demethyl-2-methoxy-24-methyl-hygrolidin Oxacyclohexadecane, hygrolidin deriv., (-)-Bafilomycin A1, NSC 381866, [7R-[3Z,5E,7R*,8S*,9S*,11E,13E,15S*,16R*[1S*,2R*,3S*(2R*,4R*,5S*,6R*)]]]-8-Hydroxy-16-[2-hydroxy-1-methyl-3-[tetrahydro-2,4-dihydroxy-5-methyl-6-(1-methylethyl)-2H-pyran-2-yl]butyl]-3,15-dimethoxy-5,7,9,11-tetramethyl-oxacyclohexadeca-3,5,11,13-tetraen-2-one
IUPAC Name (3Z,5E,7R,8S,9S,11E,13E,15S,16R)-16-[(2S,3R,4S)-4-[(2R,4R,5S,6R)-2,4-dihydroxy-5-methyl-6-propan-2-yloxan-2-yl]-3-hydroxypentan-2-yl]-8-hydroxy-3,15-dimethoxy-5,7,9,11-tetramethyl-1-oxacyclohexadeca-3,5,11,13-tetraen-2-one
SMILES OCC(=O)[C@@H](O)[C@@H](O)[C@@H](C)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C35H58O9/c1-19(2)32-24(7)27(36)18-35(40,44-32)26(9)31(38)25(8)33-28(41-10)14-12-13-20(3)15-22(5)30(37)23(6)16-21(4)17-29(42-11)34(39)43-33/h12-14,16-17,19,22-28,30-33,36-38,40H,15,18H2,1-11H3/b14-12+,20-13+,21-16+,29-17-/t22-,23+,24-,25-,26-,27+,28-,30-,31+,32+,33+,35+/m0/s1
Chemical Name or Material Bafilomycin A1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.