Learn More
Micronova™ Bag Cutter, Irradiated
Product Code. 17811922 Shop All Micronova Products
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17811922 50 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17811922 Supplier Micronova™ Supplier No. BC01IRDB
  • ONLINE EXCLUSIVE PRICE
    600.00€
    512.10€ / Pack of 1
    Save 87.90€
    14% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These Bag Cutters are supplied gamma irradiated. Disposable, with no blade changes required, they are excellent for opening raw material packages and feature a retractable safety blade guard.
  • Product Type: Bag Cutter
  • Description: Retractable Safety Blade Guard
  • Color: Orange
  • Material: Plastic
  • Sterility: Yes (Gamma Irradiated)
Specifications
Handle Material Plastic
Handle Type Ergonomic
Sterility Sterile
Color Orange
Material Plastic
Packaging Double Bagged
Quantity 50 Pk.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.